Learn More
Immunology Consultants Monoclonal Mouse anti-Human IgE Antibody Clone#4D6, 1.0 mg
SDP
Catalog No. 502414276
Encompass_Preferred
This mouse IgG2a kappa is purified using salt fractionation procedures and Protein A chromatography. It is produced from tissue culture media obtained at our facility in The United States. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with human IgE (epsilon chain specific) as determined by ELISA and western blot. It is suitable for use in blotting assays, ELISA and lateral flow. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-241-4276 Supplier Immunology Consultants Supplier No. ME80A4D6
This mouse IgG2a kappa is purified using salt fractionation procedures and Protein A chromatography. It is produced from tissue culture media obtained at our facility in The United States. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with human IgE (epsilon chain specific) as determined by ELISA and western blot. It is suitable for use in blotting assays, ELISA and lateral flow. Optimal working dilutions should be determined experimentally by the investigator.