Learn More
Bethyl Laboratories, Inc Rabbit anti-Human IgE Antibody Affinity Purified, 1mg
Supplier:  Bethyl Laboratories, Inc A80109A
Encompass_Preferred
Rabbit anti-Human IgE Antibody Affinity Purified, Reactivity Human, Applications: WB, IHC, ICC, ELISA; Host Rabbit, Antibody Type Polyclonal, Conjugate Unconjugated, Format Whole IgG
Catalog No. NC9084245