Learn More
ProteinSimple Anti-Human IgG Detection Module
Supplier:  ProteinSimple DM005
Encompass
Compatible with Jess, Abby, Wes, Peggy Sue and Sally Sue systems. Module includes: Anti-Human IgG Secondary HRP Antibody, Luminol-S, Peroxide, Antibody Diluent 2, and Streptavidin-HRP.
Catalog No. NC1382145