Learn More
Genscript Corporation Anti-Human IgG Fc 50D4 mAb
Supplier:  Genscript Corporation V9040110
Encompass
Immunoglobulin G (IgG) is a type of antibody with two antigen binding sites IgG contains two identical heavy chains and two identical light chains It constitutes 75% of serum immunoglobulins in humans IgG is further subdivided into 4 subclasses IgG1 IgG2 IgG3 and IgG4
Catalog No. 50-000-59495