Learn More
Molecular Devices Molecular Devices CloneMatrix, Methylcellulose 2.5X Concentrate, 6 x 40 mL
Catalog No. 5000014390
Encompass_Preferred
CloneMatrix 2.5X is a methylcellulose concentrate used to convert standard mammalian cell culture media into semi-solid cloning media that preserves colony position for screening and automated picking.
  • 2.5X methylcellulose concentrate for preparing semi-solid cloning media.
  • Supplied as 6 x 40 mL bottles; each 40 mL makes 100 mL final media.
  • Animal-free formulation compatible with serum-free media.
  • Supports formation of discrete clonal colonies for automated colony identification and picking.
  • Ready-to-dilute format reduces aliquoting and contamination risk.
Catalog No. 50-000-14390 Supplier Molecular Devices Supplier No. K8500
CloneMatrix 2.5X is a methylcellulose concentrate used to convert standard mammalian cell culture media into semi-solid cloning media that preserves colony position for screening and automated picking.
  • 2.5X methylcellulose concentrate for preparing semi-solid cloning media.
  • Supplied as 6 x 40 mL bottles; each 40 mL makes 100 mL final media.
  • Animal-free formulation compatible with serum-free media.
  • Supports formation of discrete clonal colonies for automated colony identification and picking.
  • Ready-to-dilute format reduces aliquoting and contamination risk.