Learn More
Seracare Life Sciences Inc ANTI-HUMAN IGG (G) AB CY5
Supplier:  Seracare Life Sciences Inc 52300368
Encompass_Preferred
Affinity purified polyclonal antibody to Human IgG heavy chains (gamma) made in Goat and labeled with Cy5. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).
Catalog No. 50-670-55