Learn More
Seracare Life Sciences Inc ANTI-HUMAN IGG (GAMMA) AB HRP
Supplier:  Seracare Life Sciences Inc 52200327
Encompass_Preferred
Affinity purified polyclonal antibody to Human IgG heavy chains (gamma) made in Goat and labeled with horseradish peroxidase. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-671-92