Learn More
Seracare Life Sciences Inc ANTI-HUMAN IGG (H+L) AB DL488
Catalog No. 5066948
Encompass_Preferred
Affinity purified polyclonal antibody to Human IgG, both heavy and light chains (whole IgG) made in Goat and labeled with DyLight 488. Product is in lyophilized form.
This item is not returnable. View return policy
Catalog No. 50-669-48 Supplier Seracare Life Sciences Inc Supplier No. 52300338
Affinity purified polyclonal antibody to Human IgG, both heavy and light chains (whole IgG) made in Goat and labeled with DyLight 488. Product is in lyophilized form.