Learn More
Seracare Life Sciences Inc ANTI-HUMAN IGG (H+L) AB FITC
Catalog No. 5066874
Encompass_Preferred
Affinity purified polyclonal antibody to Human IgG, both heavy and light chains (whole IgG) made in Goat and labeled with fluorescein isothiocyanate (FITC). Product is in lyophilized form
This item is not returnable. View return policy
Catalog No. 50-668-74 Supplier Seracare Life Sciences Inc Supplier No. 52300288
Affinity purified polyclonal antibody to Human IgG, both heavy and light chains (whole IgG) made in Goat and labeled with fluorescein isothiocyanate (FITC). Product is in lyophilized form