Learn More
Seracare Life Sciences Inc ANTI-HUMAN IGG (H+L) AB FITC
Supplier:  Seracare Life Sciences Inc 52300425
Encompass_Preferred
Affinity purified polyclonal antibody to Human IgG, both heavy and light chains (whole IgG) made in Goat and labeled with fluorescein isothiocyanate (FITC). Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-672-98