Learn More
Seracare Life Sciences Inc ANTI-HUMAN IGG (H+L) AB HRP
Supplier:  Seracare Life Sciences Inc 54500009
Encompass_Preferred
Affinity purified polyclonal antibody to Human IgG heavy and light chains (whole IgG) made in Goat and labeled with horseradish peroxidase. Product is in liquid form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-673-86