Learn More
Immunology Consultants anti-Human IgG4 Antibody (Mouse Clone #1F12) - Monoclonal
SDP
Catalog No. 5000435509
Encompass_Preferred
This mouse monoclonal antibody provides highly specific detection of human IgG4, supporting ELISA, Western blotting, and immunoassay development requiring subclass‑restricted recognition. Clone 1F12 delivers strong affinity and minimal cross‑reactivity with other IgG subclasses, enabling precise measurement of IgG4 in serum, plasma, and purified immunoglobulin preparations. Its clean, reproducible signal makes it ideal for allergy research, immune profiling, therapeutic antibody characterization, and studies examining IgG4‑associated conditions. MG4‑80A‑1F12 offers consistent lot‑to‑lot performance, ensuring reliable subclass quantification across diverse analytical workflows
Catalog No. 50-004-35509 Supplier Immunology Consultants Supplier No. MG480A1F12
This mouse monoclonal antibody provides highly specific detection of human IgG4, supporting ELISA, Western blotting, and immunoassay development requiring subclass‑restricted recognition. Clone 1F12 delivers strong affinity and minimal cross‑reactivity with other IgG subclasses, enabling precise measurement of IgG4 in serum, plasma, and purified immunoglobulin preparations. Its clean, reproducible signal makes it ideal for allergy research, immune profiling, therapeutic antibody characterization, and studies examining IgG4‑associated conditions. MG4‑80A‑1F12 offers consistent lot‑to‑lot performance, ensuring reliable subclass quantification across diverse analytical workflows