Learn More
Jackson Immuno Research Labs ANTI-HUMAN IGM
SDP
Catalog No. NC0608138
Encompass_Preferred
AffiniPure F(ab')2 Fragment Donkey Anti-Human IgM, Fc5u Fragment Specific (min X Bov,Hrs Sr Prot)
Catalog No. NC0608138 Supplier Jackson Immuno Research Labs Supplier No. 709006073
AffiniPure F(ab')2 Fragment Donkey Anti-Human IgM, Fc5u Fragment Specific (min X Bov,Hrs Sr Prot)