Learn More
Immunology Consultants Monoclonal Mouse anti-Human IgM Antibody Clone 1H12, 1.0 mg
SDP
Catalog No. 502414299
Encompass_Preferred
This mouse IgG1 kappa is purified using Protein A chromatography. It is produced from tissue culture media obtained at our facility in The United States. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with human IgM as determined by ELISA. No crossreactivity was detected with human IgG and IgA. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-241-4299 Supplier Immunology Consultants Supplier No. MM80A1H12
This mouse IgG1 kappa is purified using Protein A chromatography. It is produced from tissue culture media obtained at our facility in The United States. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with human IgM as determined by ELISA. No crossreactivity was detected with human IgG and IgA. Optimal working dilutions should be determined experimentally by the investigator.