Learn More
Immunology Consultants Monoclonal Mouse anti-Human IgM Antibody Clone 1A5, 1.0 mg
SDP
Catalog No. 502414272
Encompass_Preferred
This mouse IgG1 kappa is purified using Protein A chromatography. It is produced from tissue culture media obtained at our facility in The United States. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with human IgM as determined by ELISA and western blotting. No cross-reactivity was detected with human IgG and IgA by ELISA. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-241-4272 Supplier Immunology Consultants Supplier No. MM80A1A5
This mouse IgG1 kappa is purified using Protein A chromatography. It is produced from tissue culture media obtained at our facility in The United States. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with human IgM as determined by ELISA and western blotting. No cross-reactivity was detected with human IgG and IgA by ELISA. Optimal working dilutions should be determined experimentally by the investigator.