Learn More
Immunology Consultants Affinity Purified Goat anti-Human IgM Antibody, 1.0 mg
SDP
Catalog No. 502414275
Encompass_Preferred
Goats were immunized with highly purified human IgM and the resulting antiserum was collected. Antibody was solid phase adsorbed and immunoaffinity purified off a human IgM containing immunosorbent. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with human IgM as determined by IEP and ELISA techniques. It is suitable for blotting, ELISA and IHC applications. This antibody may cross react with IgM from other species. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-241-4275 Supplier Immunology Consultants Supplier No. GM80A
Goats were immunized with highly purified human IgM and the resulting antiserum was collected. Antibody was solid phase adsorbed and immunoaffinity purified off a human IgM containing immunosorbent. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with human IgM as determined by IEP and ELISA techniques. It is suitable for blotting, ELISA and IHC applications. This antibody may cross react with IgM from other species. Optimal working dilutions should be determined experimentally by the investigator.