Learn More
Seracare Life Sciences Inc ANTI-HUMAN IGM (MU) AB HRP
Supplier:  Seracare Life Sciences Inc 52200457
Encompass_Preferred
Affinity purified polyclonal antibody to Human IgM heavy chains (mu) made in Goat and labeled with horseradish peroxidase. Product is in liquid form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-673-85