Learn More
Rockland Immunochemicals Anti-HUMAN IgM (mu chain) (GOAT) Antibody - 609-1107
SDP
Supplier:  Rockland Immunochemicals 6091107
Encompass
Anti-Human IgM (mu heavy chain) generated in goat detects specifically Human IgM mu heavy chain. Immunoglobulin M is the largest antibody isotype and the first to be secreted against an initial exposure to antigen. IgM is predominantly produced in the spleen. Formed from covalently linking 5 immunoglobulins together. Anti-Human IgM mu chain antibody is ideal for investigators in Immunology, Microbiology, and Cell Biology.
Catalog No. 50-105-8299