Learn More
Rockland Immunochemicals Anti-HUMAN IgM (mu chain) (GOAT) Antibody DyLight™ 800 Conjugated - 609-145-007
SDP
Supplier:  Rockland Immunochemicals 609145007
Encompass
Anti-Human IgM (mu heavy chain) DyLight 800 generated in goat detects specifically Human IgM mu heavy chain. Immunoglobulin M is the largest antibody isotype and the first to be secreted against an initial exposure to antigen. IgM is predominantly produced in the spleen. Formed from covalently linking 5 immunoglobulins together. Anti-Human IgM mu chain antibody is ideal for investigators in Immunology, Microbiology, and Cell Biology.
Catalog No. 50-105-8319