Learn More
Seracare Life Sciences Inc ANTI-HUMAN IGM (U) AB CY5
Catalog No. 5067056
Encompass_Preferred
Affinity purified polyclonal antibody to Human IgM heavy chains (mu) made in Goat and labeled with Cy5. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).
This item is not returnable. View return policy
Catalog No. 50-670-56 Supplier Seracare Life Sciences Inc Supplier No. 52300369
Affinity purified polyclonal antibody to Human IgM heavy chains (mu) made in Goat and labeled with Cy5. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).