Learn More
BIOHIPPO Anti-Human IRAK4 Antibody (SAA1623), 100 µg
SDP
Catalog No. 5000860852
Encompass_Preferred
IRAK4 Recombinant Antibody (clone SAA1623) is a recombinant Mouse IgG2a, kappa antibody specific for IRAK4. It reacts with Human. Suitable for ELISA. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.
Catalog No. 50-008-60852 Supplier BIOHIPPO Supplier No. BHA21407349
IRAK4 Recombinant Antibody (clone SAA1623) is a recombinant Mouse IgG2a, kappa antibody specific for IRAK4. It reacts with Human. Suitable for ELISA. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.