Learn More
Raybiotech Inc ANTI-HUMAN MAGNETIC BEADS 4ML
Catalog No. NC2580692
Encompass_Preferred
Superparamagnetic nanoparticles coupled with Goat anti-Human IgG and utilized in the magnetic separation and isolation of human antibodies from serum or human antibody-labeled components.
Catalog No. NC2580692 Supplier Raybiotech Inc Supplier No. 8011014
Superparamagnetic nanoparticles coupled with Goat anti-Human IgG and utilized in the magnetic separation and isolation of human antibodies from serum or human antibody-labeled components.