Learn More
BIOHIPPO Anti-Human MET/c-Met/HGFR Antibody (SAA0765), 100 µg
SDP
Catalog No. 5000861241
Encompass_Preferred
MET/c-Met/HGFR Recombinant Antibody (clone SAA0765) is a recombinant Human IgG1 antibody specific for MET/c-Met/HGFR. It reacts with Human. Suitable for ELISA, WB. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.
Catalog No. 50-008-61241 Supplier BIOHIPPO Supplier No. BHA21408295
MET/c-Met/HGFR Recombinant Antibody (clone SAA0765) is a recombinant Human IgG1 antibody specific for MET/c-Met/HGFR. It reacts with Human. Suitable for ELISA, WB. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.