Learn More
Medchemexpress LLC Anti-Human Mouse den 5mg | 5mg
SDP
Catalog No. 5000325813
Encompass_Preferred
Anti-Human/Mouse denatured collagen type-I Antibody (XL313) is a mouse-derived IgG1 type antibody inhibitor targeting to human/mouse denatured collagen type-I Anti-Human/Mouse denatured collagen type-I Antibody (XL313) selectively binds to proteolyzed collagen type I Anti-Human/Mouse denatured collagen type-I Antibody (XL313) reduces PD L1 levels in tumor cells Anti-Human/Mouse denatured collagen type-I Antibody (XL313) can be used for the researches of cancer and inflammation such as such as ovarian tumor[1][2][3]
Catalog No. 50-003-25813 Supplier Medchemexpress LLC Supplier No. HYP9902515MG
Anti-Human/Mouse denatured collagen type-I Antibody (XL313) is a mouse-derived IgG1 type antibody inhibitor targeting to human/mouse denatured collagen type-I Anti-Human/Mouse denatured collagen type-I Antibody (XL313) selectively binds to proteolyzed collagen type I Anti-Human/Mouse denatured collagen type-I Antibody (XL313) reduces PD L1 levels in tumor cells Anti-Human/Mouse denatured collagen type-I Antibody (XL313) can be used for the researches of cancer and inflammation such as such as ovarian tumor[1][2][3]