Learn More
Selleck Chemical LLC Anti-human/mouse TGF-b-InVivo
SDP
Supplier:  Selleck Chemical LLC A21135MG 5PC
Encompass_Preferred
The 1D11 16 8 monoclonal antibody reacts with mouse human rat monkey hamster canine and bovine TGF- (transforming growth factor beta) isoforms 1 2 and 3 The 1D11 16 8 monoclonal antibody is a neutralizing antibody
Catalog No. 50-313-6552