Learn More
Selleck Chemical LLC Anti-human/mouse TGF-b-InVivo
SDP
Catalog No. 503135909
Encompass_Preferred
The 1D11 16 8 monoclonal antibody reacts with mouse human rat monkey hamster canine and bovine TGF- (transforming growth factor beta) isoforms 1 2 and 3 The 1D11 16 8 monoclonal antibody is a neutralizing antibody
Catalog No. 50-313-5909 Supplier Selleck Chemical LLC Supplier No. A21135MG 20PC
The 1D11 16 8 monoclonal antibody reacts with mouse human rat monkey hamster canine and bovine TGF- (transforming growth factor beta) isoforms 1 2 and 3 The 1D11 16 8 monoclonal antibody is a neutralizing antibody