Learn More
Selleck Chemical LLC Anti-human PD-L1-InVivo
SDP
Supplier:  Selleck Chemical LLC A21561MG
Encompass_Preferred
Anti-human PD-L1-InVivo reacts with human PD-L1 (programmed death ligand 1) also known as B7-H1 or CD274 PD-L1 is a 40 kDa type I transmembrane protein that belongs to the B7 family of the Ig superfamily
Catalog No. 50-313-8070