Learn More
BIOHIPPO Anti-Human PG I/PGA Monoclonal Antibody (1A287), 100 µg
SDP
Catalog No. 5000857979
Encompass_Preferred
Human PG I/PGA Monoclonal Antibody is a Mouse-derived monoclonal antibody that specifically recognizes PG I/PGA. It shows reactivity with Human. Validated applications include ELISA, TIA. Protein A/G purified from cell culture supernatant. Supplied as 100 ug.
Catalog No. 50-008-57979 Supplier BIOHIPPO Supplier No. BHA21416114
Human PG I/PGA Monoclonal Antibody is a Mouse-derived monoclonal antibody that specifically recognizes PG I/PGA. It shows reactivity with Human. Validated applications include ELISA, TIA. Protein A/G purified from cell culture supernatant. Supplied as 100 ug.