Learn More
BIOHIPPO Anti-Human PG I/PGA Monoclonal Antibody (1A288), 100 µg
SDP
Catalog No. 5000858330
Encompass_Preferred
Human PG I/PGA Monoclonal Antibody is a Mouse-derived monoclonal antibody that specifically recognizes PG I/PGA. It shows reactivity with Human. Validated applications include IFA, TIA. Protein A/G purified from cell culture supernatant. Supplied as 100 ug.
Catalog No. 50-008-58330 Supplier BIOHIPPO Supplier No. BHA21416113
Human PG I/PGA Monoclonal Antibody is a Mouse-derived monoclonal antibody that specifically recognizes PG I/PGA. It shows reactivity with Human. Validated applications include IFA, TIA. Protein A/G purified from cell culture supernatant. Supplied as 100 ug.