Learn More
BIOHIPPO Anti-Human PG II/PGC Monoclonal Antibody (1A290), 100 µg
SDP
Catalog No. 5000857233
Encompass_Preferred
Human PG II/PGC Monoclonal Antibody is a Mouse-derived monoclonal antibody that specifically recognizes PG II/PGC. It shows reactivity with Human. Validated applications include ELISA, IFA, TIA. Protein A/G purified from cell culture supernatant. Supplied as 100 ug.
Catalog No. 50-008-57233 Supplier BIOHIPPO Supplier No. BHA21416111
Human PG II/PGC Monoclonal Antibody is a Mouse-derived monoclonal antibody that specifically recognizes PG II/PGC. It shows reactivity with Human. Validated applications include ELISA, IFA, TIA. Protein A/G purified from cell culture supernatant. Supplied as 100 ug.