Learn More
BIOHIPPO Anti-Human PTK7/CCK-4 Antibody (SAA0144), APC, 100 Tests
SDP
Catalog No. 5000859269
Encompass_Preferred
Human PTK7/CCK-4 Antibody (clone SAA0144) is a APC-conjugated monoclonal antibody for flow cytometry and related applications. It detects PTK7/CCK-4 in Human samples. Validated applications include ELISA, FCM. Supplied as 100 tests.
Catalog No. 50-008-59269 Supplier BIOHIPPO Supplier No. BHA21410786
Add to Cart
Add to Cart
Human PTK7/CCK-4 Antibody (clone SAA0144) is a APC-conjugated monoclonal antibody for flow cytometry and related applications. It detects PTK7/CCK-4 in Human samples. Validated applications include ELISA, FCM. Supplied as 100 tests.