Learn More
BIOHIPPO Anti-Human PTK7/CCK-4 Antibody (SAA0144), PerCP, 100 Tests
SDP
Catalog No. 5000856591
Encompass_Preferred
Human PTK7/CCK-4 Antibody (clone SAA0144) is a PerCP-conjugated monoclonal antibody for flow cytometry and related applications. It detects PTK7/CCK-4 in Human samples. Validated applications include ELISA, FCM. Supplied as 100 tests.
Catalog No. 50-008-56591 Supplier BIOHIPPO Supplier No. BHA21419838
Add to Cart
Add to Cart
Human PTK7/CCK-4 Antibody (clone SAA0144) is a PerCP-conjugated monoclonal antibody for flow cytometry and related applications. It detects PTK7/CCK-4 in Human samples. Validated applications include ELISA, FCM. Supplied as 100 tests.