Learn More
Medchemexpress LLC Anti-Human Rat Fish 5mg | 5mg
SDP
Catalog No. 5000325441
Encompass_Preferred
Anti-Human/Rat/Fish AChR Antibody (Mab35) is an anti-human/rat/fish AChR IgG1 monoclonal antibody Anti-Human/Rat/Fish AChR Antibody (Mab35) can attack the AChR of neuromuscular junctions Anti-Human/Rat/Fish AChR Antibody (Mab35) can be used for the construction of myasthenia gravis models[1][2][3]
Catalog No. 50-003-25441 Supplier Medchemexpress LLC Supplier No. HYP9903025MG
Anti-Human/Rat/Fish AChR Antibody (Mab35) is an anti-human/rat/fish AChR IgG1 monoclonal antibody Anti-Human/Rat/Fish AChR Antibody (Mab35) can attack the AChR of neuromuscular junctions Anti-Human/Rat/Fish AChR Antibody (Mab35) can be used for the construction of myasthenia gravis models[1][2][3]