Learn More
Medchemexpress LLC Anti-Human Rat HER2 10mg | 10mg
SDP
Catalog No. 5000324475
Encompass_Preferred
Anti-Human/Rat HER2 (neu) Antibody (7 16 4) is an anti-mouse HER2 IgG2a monoclonal antibody Anti-Human/Rat HER2 (neu) Antibody (7 16 4) reduces tumor cell proliferation by inhibiting the HER2 signaling pathway Anti-Human/Rat HER2 (neu) Antibody (7 16 4) can activate CD8 T cells Anti-Human/Rat HER2 (neu) Antibody (7 16 4) can be used for research on cancer such as breast cancer[1][2]
Catalog No. 50-003-24475 Supplier Medchemexpress LLC Supplier No. HYP99023810MG
Anti-Human/Rat HER2 (neu) Antibody (7 16 4) is an anti-mouse HER2 IgG2a monoclonal antibody Anti-Human/Rat HER2 (neu) Antibody (7 16 4) reduces tumor cell proliferation by inhibiting the HER2 signaling pathway Anti-Human/Rat HER2 (neu) Antibody (7 16 4) can activate CD8 T cells Anti-Human/Rat HER2 (neu) Antibody (7 16 4) can be used for research on cancer such as breast cancer[1][2]