Learn More
Medchemexpress LLC Anti-Human Rat HER2 5mg | 5mg
SDP
Catalog No. 5000324474
Encompass_Preferred
Anti-Human/Rat HER2 (neu) Antibody (7 16 4) is an anti-mouse HER2 IgG2a monoclonal antibody Anti-Human/Rat HER2 (neu) Antibody (7 16 4) reduces tumor cell proliferation by inhibiting the HER2 signaling pathway Anti-Human/Rat HER2 (neu) Antibody (7 16 4) can activate CD8 T cells Anti-Human/Rat HER2 (neu) Antibody (7 16 4) can be used for research on cancer such as breast cancer[1][2]
Catalog No. 50-003-24474 Supplier Medchemexpress LLC Supplier No. HYP9902385MG
Anti-Human/Rat HER2 (neu) Antibody (7 16 4) is an anti-mouse HER2 IgG2a monoclonal antibody Anti-Human/Rat HER2 (neu) Antibody (7 16 4) reduces tumor cell proliferation by inhibiting the HER2 signaling pathway Anti-Human/Rat HER2 (neu) Antibody (7 16 4) can activate CD8 T cells Anti-Human/Rat HER2 (neu) Antibody (7 16 4) can be used for research on cancer such as breast cancer[1][2]