Learn More
Mabtech Inc Anti-human IgG mAbs MT91/145, purified, 1 mg
Supplier:  Mabtech Inc 385031000
Encompass
These monoclonal antibodies enable sensitive and specific detection of human IgG in immunoassays such as ELISpot.
Catalog No. NC0306931