Learn More
Biolegend APC anti-human TGF-β Receptor II
Supplier:  Biolegend 399705
Encompass_Preferred
APC anti-human TGF-β Receptor II [W17055E]; Isotype: Rat IgG2b, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. NC1919939