Learn More
Quidel Corporation HUMAN SC5B-9 MAB BIOTIN
SDP
Supplier:  Quidel Corporation A711
Encompass
A biotin conjugated murine monoclonal antibody to a neoantigen on the terminal complement complex.
Catalog No. NC1887683