Learn More
STEMCELL Technologies Anti-Human SSEA-4 Antibody, Clone MC-813-70, PE, Size: 25 Tests
SDP
Supplier:  STEMCELL Technologies 60062PE.1
Encompass
Mouse monoclonal IgG3 antibody against human, mouse, rat SSEA-4, PE-conjugated
Catalog No. 50-197-5335