Learn More
STEMCELL Technologies Anti-Human SSEA-5 Antibody, Clone 8e11, Size: 50 µg
SDP
Supplier:  STEMCELL Technologies 60063.1
Encompass
Clone 8e11 is a mouse monoclonal antibody that reacts with human stage-specific embryonic antigen-5 (SSEA-5), a terminal H type-1 glycan (Fuc1-2Galβ1-3GlcNAcβ1) expressed on the surface of cells in the inner cell mass of the human blastocyst during embryogenesis, and on undifferentiated human embryonic stem (ES) and induced pluripotent stem (iPS) cells. Verified for use with flow cytometry, immunocytochemistry, immunofluorescence microscopy, and western blotting assays, use this antibody in applications requiring the detection of SSEA-5. The 8e11 antibody can be used in conjunction with other pluripotency surface markers such as CD9 and CD90, or CD50 and CD200, to sort and remove undifferentiated teratoma-forming cells from incompletely differentiated stem cell cultures. Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5337