Learn More
STEMCELL Technologies Anti-Human TRA-1-81 Antibody, Clone TRA-1-81, APC, Size: 100 Tests
SDP
Supplier:  STEMCELL Technologies 60065AZ
Encompass
Clone TRA-1-81 is a mouse monoclonal antibody that reacts with human, rat, and rhesus TRA­1­81, a high-molecular-mass (> 200 kDa) glycoprotein expressed on the surface of human embryonal carcinoma (EC), embryonic germ (EG), embryonic stem (ES), and induced pluripotent stem (iPS) cells, as well as rhesus monkey and rat ES cell lines. Verified for use with flow cytometry, immunocytochemistry, immunofluorescence microscopy, and western blotting assays, use this antibody in applications requiring the detection of TRA-1-81. The epitope of the TRA-1-81 antibody is associated with a keratan-sulfated transmembrane protein identified as podocalyxin and is resistant to treatment with neuraminidase. Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5352