Learn More
Abcam Anti-Iba1 antibody [EPR16588] – Chicken IgY (Chimeric)
Supplier:  Abcam AB31830220UL
Encompass_Preferred
Chicken Recombinant Monoclonal Iba1 antibody EPR16588. Suitable for IHC-P, ICC/IF, IHC-Fr and reacts with Human, Mouse, Rat samples.
Catalog No. NC3790583