Learn More
FUJIFILM BIOSCIENCES INC Anti Iba1, Rabbit(for Paraffin Section) | 50ug
Supplier:  FUJIFILM BIOSCIENCES INC 01327691
Encompass_Preferred
Iba1 is a protein of about 17 kDa that is expressed specifically in microglia of the nervous system, and is frequently used as a microglial marker. This product is a rabbit polyclonal antibody that recognizes Iba1. It is for immunohistological staining with paraffin section.
Catalog No. NC1718288