Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IgG, IgA, IgM, Polyclonal, FITC, MP Biomedicals

Catalog No. ICN55499 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN55499 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN55499 Supplier MP Biomedicals, Inc Supplier No. 0855499
Only null left
Add to Cart
Add to Cart

  • Conjugate: FITC
  • Host species: Goat
  • Species reactivity: Mouse

Immunoassays

TRUSTED_SUSTAINABILITY

Specifications

Conjugate FITC
Purification Method Affinity Purified
Quantity 2 mL
Primary or Secondary Secondary
Target Species Mouse
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.