Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IgG1, AmCyan, Clone: X40, BD

Catalog No. BDB339185 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB339185 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB339185 Supplier BD Biosciences Supplier No. 339185
Add to Cart
Add to Cart

Host Species: Mouse
Clone: X40
Isotype: IgG1, κ
Cross Reactivity: Human

Control, Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Concentration 100μg/mL
Conjugate AmCyan
Formulation Phosphate buffered saline with BSA and 0.1% sodium azide.
Host Species Mouse
Immunogen Keyhole limpet hemocyanin (KLH)
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Content And Storage Store vials at 2°C-8°C. Conjugated forms should not be frozen. Protect from exposure to light. Each reagent is stable until the expiration date shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.