Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IgG1, κ, Mouse, Alexa Fluor™ 488, Clone: MOPC-21, BD

Catalog No. BDB557721 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
50 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557721 100 Tests
BDB557782 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB557721 Supplier BD Biosciences Supplier No. 557721
Add to Cart
Add to Cart

The MOPC-21 immunoglobulin is a mouse myeloma protein. The MOPC-21 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: MOPC-21
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry, Intracellular Staining, Isotype Control

TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry, Western Blot
Clone MOPC-21
Conjugate Alexa Fluor 488
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Target Species Human
Content And Storage Undiluted at 4°C
Isotype IgG1 κ
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.