Learn More
Description
The G155-178 clone has an unknown specificity. Trinitrophenol (TNP), the immunogen, is a hapten not expressed on human, mouse, rat or non-human primate cells. In the absence of specific binding, this antibody may bind non-specifically to immunoglobulin Fc receptors. The immunoglobulin secreted by the G155-178 hybridoma was selected as an mouse IgG2a, ? isotype control following screening for low background binding on a variety of mouse and human tissues. The antibody was conjugated to Brilliant Violet 421 and has been developed in collaboration with Sirigen. With an Ex Max of 407-nm and Em Max at 421-nm, Brilliant Violet 421 can be excited by the violet laser and detected in the standard Pacific Blue filter set (eg,
450/50-nm filter). Brilliant Violet 421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.
Host Species: Mouse
Clone: G155-178
Isotype: IgG2a, κ
Immunogen: TNP-keyhole limpet hemocyanin
Flow Cytometry, Isotype Control
Specifications
Specifications
| Applications | Flow Cytometry, Immunocytology |
| Clone | G155-178 |
| Conjugate | Brilliant Violet 421 |
| Host Species | Mouse |
| Immunogen | TNP-keyhole limpet hemocyanin |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Content And Storage | 4°C, protect from prolonged exposure to light, DO NOT FREEZE |
| Isotype | IgG2a κ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.