Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IgG2b κ, Mouse , Clone: MPC-11, BD

Catalog No. BDB557351 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg/mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557351 0.5 mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557351 Supplier BD Biosciences Supplier No. 557351
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MPC-11 antibody has unknown specificity. The MPC-11 cell line was adapted to continuous culture from the tranplantable Merwin Plasmacytoma-11, which was induced by intraperitoneal implantation of a Millipore diffusion chamber into a BALB/c mouse. This antibody is routinely tested by ELISA, immunohistochemical and flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: MPC-11
Isotype: IgG2b, κ

Cytotoxicity Assay, ELISA Standard, Flow Cytometry, Immunohistochemistry (Formalin-Antigen Retrieval Required), Immunohistochemistry (Frozen), Isotype Control

TRUSTED_SUSTAINABILITY

Specifications

Applications Cytotoxicity Assay, ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Clone MPC-11
Concentration 0.5mg/mL
Conjugate Unconjugated
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.5 mg/mL
Content And Storage Undiluted at 4°C
Isotype IgG2b κ
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.