Learn More
Sino Biological Anti-IL-1 alpha/IL1A Antibody (Biotin), Rabbit Polyclonal 100 µg
Catalog No. 5000749049
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL-1 alpha / IL-1F1 / IL1A (Catalog#50114-MNAE; NP_034684.2; Ser115-Ser270). Total IgG was purified by Protein A affinity chromatographyand then biotinylated.
Catalog No. 50-007-49049 Supplier Sino Biological Supplier No. 50114RP01B100
Produced in rabbits immunized with purified, recombinant Mouse IL-1 alpha / IL-1F1 / IL1A (Catalog#50114-MNAE; NP_034684.2; Ser115-Ser270). Total IgG was purified by Protein A affinity chromatographyand then biotinylated.