Learn More
Sino Biological Anti-IL-1 alpha/IL1A Antibody, Rabbit Polyclonal 100
Catalog No. 5000750444
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL-1 alpha/IL1A (rh IL-1 alpha/IL1A; Catalog#10128-HNCH; NP_000566.3; Ser113-Ala271). IL-1 alpha/IL1A specific IgG was purified by Human IL-1 alpha/IL1A affinity chromatography.
Catalog No. 50-007-50444 Supplier Sino Biological Supplier No. 10128T48100
Produced in rabbits immunized with purified, recombinant Human IL-1 alpha/IL1A (rh IL-1 alpha/IL1A; Catalog#10128-HNCH; NP_000566.3; Ser113-Ala271). IL-1 alpha/IL1A specific IgG was purified by Human IL-1 alpha/IL1A affinity chromatography.